
tert-Butyl-3-aminobenzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:92146-82-2
:
Formula:C₁₁H₁₅NO₂
MDL Number:MFCD00729048
:193.2423 g/mol
Melting Pt:76…78 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together