
2-Amino-5-ethyl-1,3,4-thiadiazole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:14068-53-2
:
Formula:C₄H₇N₃S
MDL Number:MFCD00003111
:129.19 g/mol
Melting Pt:198…202 °C
EINECS:237-921-4
:GHS07
Storage Temperature:Ambient
Frequently Bought Together