Order Entry
Czech Republic
ContactUsLinkComponent
2-Bromo-1-(4-chlorophenyl)-2-(4-methylphenyl)ethanone
Corrosive
2-Bromo-1-(4-chlorophenyl)-2-(4-methylphenyl)ethanone
Catalog # APOSOR24072-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
2-Bromo-1-(4-chlorophenyl)-2-(4-methylphenyl)ethanone
Catalog # APOSOR24072-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :323.62 g/mol
Melting Pt:76…78 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
 :GHS05
CAS Number:119267-79-7
Formula:C₁₅H₁₂BrClO
MDL Number:MFCD00052202