Order Entry
Czech Republic
ContactUsLinkComponent
 
Irritant
5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulphonamide
Catalog # APOSOR23929-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-Chloro-1,3-dimethyl-1H-pyrazole-4-sulphonamide
Catalog # APOSOR23929-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :209.66 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:88398-46-3
Formula:C₅H₈ClN₃O₂S
MDL Number:MFCD00105874