Order Entry
Czech Republic
ContactUsLinkComponent
 
Ethyl-2-amino-4-methylpyrimidine-5-carboxylate 97%
Catalog # APOSOR23898-1G
Supplier:  Apollo Scientific
CAS Number:  
Ethyl-2-amino-4-methylpyrimidine-5-carboxylate 97%
Catalog # APOSOR23898-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :181.2 g/mol
Melting Pt:220 °C
Storage Temperature:Ambient
CAS Number:81633-29-6
Formula:C₈H₁₁N₃O₂
MDL Number:MFCD00052622