
6-Morpholino-3-pyridinamine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:52023-68-4
:
Formula:C₉H₁₃N₃O
MDL Number:MFCD00119562
:179.22 g/mol
Melting Pt:126…130 °C
Storage Temperature:Ambient
EINECS:257-613-3
UN:2811
ADR:6.1,III
:GHS07
Frequently Bought Together