Order Entry
Czech Republic
ContactUsLinkComponent
4-(3-Aminopropyl)-2,4-dihydro-3H-pyrazol-3-one
Corrosive
4-(3-Aminopropyl)-2,4-dihydro-3H-pyrazol-3-one
Catalog # APOSOR23568-250MG
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
4-(3-Aminopropyl)-2,4-dihydro-3H-pyrazol-3-one
Catalog # APOSOR23568-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :141.17 g/mol
Melting Pt:216 °C
Storage Temperature:Ambient
UN:3259
ADR:8,III
 :GHS05
CAS Number:7032-17-9
Formula:C₆H₁₁N₃O
MDL Number:MFCD00053049