

1-[3-(Chloromethyl)-4-methoxyphenyl]ethanone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:62581-82-2
:
Formula:C₁₀H₁₁ClO₂
MDL Number:MFCD00045267
:198.65 g/mol
Melting Pt:116…118 °C
UN:3261
ADR:8,III
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together