
1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:85233-19-8
:
Formula:C₂₂H₂₄N₂O₁₀
MDL Number:MFCD00036255
:476.44 g/mol
Melting Pt:189…190 °C
:GHS07
Storage Temperature:Freezer
Frequently Bought Together