Order Entry
Czech Republic
ContactUsLinkComponent
N-(4-Methyl-1,3-thiazol-2-yl)guanidine hydrochloride
Irritant
N-(4-Methyl-1,3-thiazol-2-yl)guanidine hydrochloride
Catalog # APOSOR23493-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
N-(4-Methyl-1,3-thiazol-2-yl)guanidine hydrochloride
Catalog # APOSOR23493-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :192.67 g/mol
Melting Pt:188…190 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:100599-91-5
Formula:C₅H₉ClN₄S
MDL Number:MFCD00053024