0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Bis(4-methoxyphenyl)phenylphosphine
Bis(4-methoxyphenyl)phenylphosphine
Bis(4-methoxyphenyl)phenylphosphine
APOSOR23446-5G
:
Apollo Scientific
:
Bis(4-methoxyphenyl)phenylphosphine
APOSOR23446-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
322.34 g/mol
:
88…90 °C
:
GHS07
:
Ambient
:
14180-51-9
:
C₂₀H₁₉O₂P
:
MFCD00048993