
(5-Methylisoxazol-3-yl)methylamine hydrochloride
Catalog # APOSOR23144-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:148.59 g/mol
Boiling Pt:75…76 °C/0.4mmHg
Storage Temperature:Ambient
UN:2735
ADR:8,III
:GHS05
CAS Number:154016-48-5
Formula:C₅H₉ClN₂O
MDL Number:MFCD02681995