
4-Bromo-3,5-dimethyl-1H-pyrazole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3398-16-1
:
Formula:C₅H₇BrN₂
MDL Number:MFCD00005242
:175.03 g/mol
Melting Pt:123…125 °C
EINECS:222-263-2
:GHS07
Storage Temperature:Ambient
Frequently Bought Together