
4-Cyclohexylbenzenesulphonyl chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:56354-57-5
:
Formula:C₁₂H₁₅ClO₂S
MDL Number:MFCD00625713
:258.76 g/mol
Melting Pt:42…52 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:GHS05
Frequently Bought Together