1-[2-Hydroxy-6-(isopentyloxy)phenyl]ethan-1-one
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:249278-25-9
:
Formula:C₁₃H₁₈O₃
MDL Number:MFCD00100622
:222.29 g/mol
Melting Pt:25…27 °C
Storage Temperature:Ambient
Frequently Bought Together