Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-{4-[(3,4-Dichlorobenzyl)oxy]phenyl}-1-ethanone
1-{4-[(3,4-Dichlorobenzyl)oxy]phenyl}-1-ethanone
1-{4-[(3,4-Dichlorobenzyl)oxy]phenyl}-1-ethanone
1-{4-[(3,4-Dichlorobenzyl)oxy]phenyl}-1-ethanone
APOSOR21843-250MG
:
Apollo Scientific
:
1-{4-[(3,4-Dichlorobenzyl)oxy]phenyl}-1-ethanone
APOSOR21843-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
295.17 g/mol
:
83…85 °C
:
GHS07
:
Ambient
:
170916-55-9
:
C₁₅H₁₂Cl₂O₂
:
MFCD00173669