
4,5-Dichloro-2-methyl-2H-pyridazin-3-one
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:933-76-6
:
Formula:C₅H₄Cl₂N₂O
MDL Number:MFCD00051686
Melting Pt:87…91 °C
UN:2811
ADR:6.1,III
:179.01 g/mol
:GHS06
Storage Temperature:Ambient
Frequently Bought Together