
4-Chloro-2-methyl-5-(1-methylhydrazino)-2,3-dihydropyridazin-3-one ≥90%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:96017-23-1
:
Formula:C₆H₉ClN₄O
MDL Number:MFCD00052906
:188.62 g/mol
Melting Pt:131…133 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together