
6-Chloro-4-nitro-o-cresol
Catalog # APOSOR21328-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:GHS07
Storage Temperature:Ambient
CAS Number:4102-84-5
Formula:C₇H₆ClNO₃
MDL Number:MFCD00047740