
5-Bromo-2-methoxybenzenesulphonyl chloride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:23095-05-8
:
Formula:C₇H₆BrClO₃S
MDL Number:MFCD00051768
:285.55 g/mol
Melting Pt:115…118 °C
Storage Temperature:Refrigerator
UN:3261
ADR:8,II
:GHS05
Frequently Bought Together