
Pyridine-2-carboxaldoxime
Catalog # APOSOR2013-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:122.13 g/mol
Melting Pt:110…112 °C
:GHS07
Storage Temperature:Ambient
CAS Number:873-69-8
Formula:C₆H₆N₂O
MDL Number:MFCD00006354