Order Entry
Czech Republic
ContactUsLinkComponent
 
5-Nitro-1,3-dihydro-2H-benzimidazol-2-one 97%
Catalog # APOSOR200107-25G
Supplier:  Apollo Scientific
CAS Number:  
5-Nitro-1,3-dihydro-2H-benzimidazol-2-one 97%
Catalog # APOSOR200107-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :179.14 g/mol
Storage Temperature:Ambient
CAS Number:93-84-5
Formula:C₇H₅N₃O₃
MDL Number:MFCD00220274