N-(2,4-Dinitrophenyl)-1-methylpiperidin-4-amine
Catalog # APOSOR200039-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:280.29 g/mol
Storage Temperature:Ambient
CAS Number:925216-76-8
Formula:C₁₂H₁₆N₄O₄
MDL Number:MFCD22126063