
Methyl-3-(pyrrolidin-1-ylmethyl)benzoate
Catalog # APOSOR200036-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:219.29 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:321198-22-5
Formula:C₁₃H₁₇NO₂
MDL Number:MFCD09909459