Methyl-3-[(1,3-dioxo-1,3-dihydro-2H-isoindol-2-yl)methyl]benzoate
Catalog # APOSOR200009-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:295.3 g/mol
Storage Temperature:Ambient
CAS Number:781632-38-0
Formula:C₁₇H₁₃NO₄
MDL Number:MFCD06353849