Order Entry
Czech Republic
ContactUsLinkComponent
Methyl-2-{[(4-methoxybenzyl)amino]methyl}benzoate
Methyl-2-{[(4-methoxybenzyl)amino]methyl}benzoate
Catalog # APOSOR200007-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl-2-{[(4-methoxybenzyl)amino]methyl}benzoate
Catalog # APOSOR200007-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :285.35 g/mol
Storage Temperature:Ambient
CAS Number:1157652-59-9
Formula:C₁₇H₁₉NO₃
MDL Number:MFCD13790489