
6-Hydroxypyrimidine-4-carboxylic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:6299-87-2
:
Formula:C₅H₄N₂O₃
MDL Number:MFCD00023267
:140.0969 g/mol
Melting Pt:276 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together