
2-Methoxyisonicotinic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:105596-63-2
:
Formula:C₇H₇NO₃
MDL Number:MFCD04115718
:153.1400 g/mol
Melting Pt:197…200 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together