Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Registrovat
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Nitro-1H-pyrazole
3-Nitro-1H-pyrazole
3-Nitro-1H-pyrazole
3-Nitro-1H-pyrazole
APOSOR18620-25G
:
Apollo Scientific
:
3-Nitro-1H-pyrazole
APOSOR18620-25G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
25 g
Seq:
3308
:
:
113.08 g/mol
:
173…175 °C
:
GHS07
:
Ambient
:
26621-44-3
:
C₃H₃N₃O₂
:
MFCD00238787