


(R)-2-Methylpyrrolidine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:135324-85-5
:
Formula:C₅H₁₂ClN
MDL Number:MFCD06796610
ADR:4.1+8,II
:121.6085 g/mol
UN:2925
:GHS02
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together