
2-(4-Hydroxyphenylazo)benzoic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1634-82-8
:
Formula:C₁₃H₁₀N₂O₃
MDL Number:MFCD00002428
:242.23 g/mol
Melting Pt:204…208 °C
EINECS:216-655-2
:GHS07
Storage Temperature:Ambient
Frequently Bought Together