Order Entry
Czech Republic
ContactUsLinkComponent
2-(1H-Pyrazol-5-yl)aniline
2-(1H-Pyrazol-5-yl)aniline
Catalog # APOSOR183453-250MG
Supplier:  Apollo Scientific
CAS Number:  
2-(1H-Pyrazol-5-yl)aniline
Catalog # APOSOR183453-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :159.1879 g/mol
Melting Pt:129 °C
Storage Temperature:Ambient
CAS Number:111562-32-4
Formula:C₉H₉N₃
MDL Number:MFCD09028476