1,1'-Carbonyldiimidazole 98%
: Di(1H-imidazol-1-yl)methanone, CDI Crosslinker, 1,1'-Carbonylbis(imidazole), 1,1'-Carbonylbis-1H-imidazole, N,N'-Carbonyldiimidazole, CDI
:530-62-1
:
:C₇H₆N₄O
:MFCD00005286
:162.15 g/mol
:117…122 °C
:8 II
:14,01819
:208-488-9
:3263
:GHS05
:GHS07
:Ambient
- Environmentally Preferable:
Frequently Bought Together