Order Entry
Czech Republic
ContactUsLinkComponent
1-(2-Hydroxyethyl)piperazine 97%
Corrosive
Irritant
1-(2-Hydroxyethyl)piperazine 97%
Catalog # APOSOR18310-500G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
1-(2-Hydroxyethyl)piperazine 97%
Catalog # APOSOR18310-500G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :130.19 g/mol
Boiling Pt:242…246 °C
Melting Pt:–39 °C
Density:1.061 g/cm³ (25 °C)
Flash Pt:135 °C
EINECS:203-142-3
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:103-76-4
Formula:C₆H₁₄N₂O
MDL Number:MFCD00005970