
2,6-Dimethylisonicotinonitrile 97%
Catalog # APOSOR18034-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:132.16 g/mol
Melting Pt:212…216 °C
:GHS07
Storage Temperature:Ambient
CAS Number:39965-81-6
Formula:C₈H₈N₂
MDL Number:MFCD00152363