


Bis(cyclopentadienyl)titanium dichloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1271-19-8
:
Formula:C₁₀H₁₀Cl₂Ti
MDL Number:MFCD00003723
:248.97 g/mol
Melting Pt:260…280 °C (dec.)
Density:1.6 g/cm³ (25 °C)
EINECS:215-035-9
UN:3261
ADR:8,II
:GHS05
:GHS07
:GHS08
Storage Temperature:Ambient
Frequently Bought Together