


Cyclopropanecarbonyl chloride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:4023-34-1
:
Formula:C₄H₅ClO
MDL Number:MFCD00001277
:104.54 g/mol
Boiling Pt:119…121 °C
Density:1.152 g/cm³ (25 °C)
Flash Pt:23 °C
EINECS:223-684-4
UN:3286
ADR:3+6.1+8,II
:GHS02
:GHS05
:GHS06
Storage Temperature:Refrigerator
Frequently Bought Together