
Methyl-3-bromo-5-methoxybenzoate
Catalog # APOSOR17115-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Boiling Pt:156…157 °C/4mmHg
:GHS07
Storage Temperature:Ambient
CAS Number:56709-70-7
Formula:C₉H₉BrO₃
MDL Number:MFCD11042779