Order Entry
Czech Republic
ContactUsLinkComponent
2-Methylsulphanyl-7H-pyrrolo[2,3-d]pyrimidin-4-ol
Irritant
2-Methylsulphanyl-7H-pyrrolo[2,3-d]pyrimidin-4-ol
Catalog # APOSOR17056-500MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Methylsulphanyl-7H-pyrrolo[2,3-d]pyrimidin-4-ol
Catalog # APOSOR17056-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :181.22 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:67831-83-8
Formula:C₇H₇N₃OS
MDL Number:MFCD02668244