
5-Pyridin-3-yl-1H-pyrazole-3-carboxylic acid
Catalog # APOSOR17049-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:189.18 g/mol
Melting Pt:308…310 °C
:GHS07
Storage Temperature:Ambient
CAS Number:374064-01-4
Formula:C₉H₇N₃O₂
MDL Number:MFCD05170031