




Allyl bromide 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:106-95-6
:
Formula:C₃H₅Br
MDL Number:MFCD00000244
:120.98 g/mol
Boiling Pt:70…71 °C
Melting Pt:–119 °C
Density:1.398 g/cm³ (25 °C)
Flash Pt:–1 °C
EINECS:203-446-6
UN:1099
ADR:3+6.1,I
:GHS02
:GHS05
:GHS06
:GHS08
:GHS09
Storage Temperature:Refrigerator
Frequently Bought Together