
tert-Butyl-2,8-diazaspiro[4.5]decane-2-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:336191-17-4
:
Formula:C₁₃H₂₄N₂O₂
MDL Number:MFCD06739056
:240.3419 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together