Order Entry
Czech Republic
ContactUsLinkComponent
 
Corrosive
Irritant
3-(2-Methyl-1,3-dioxolan-2-yl)-1-propanamine
Catalog # APOSOR16878-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
3-(2-Methyl-1,3-dioxolan-2-yl)-1-propanamine
Catalog # APOSOR16878-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :145.2 g/mol
Boiling Pt:91…92 °C/14mmHg
Storage Temperature:Ambient
UN:2735
ADR:8,III
 :GHS05
 :GHS07
CAS Number:66442-97-5
Formula:C₇H₁₅NO₂
MDL Number:MFCD09923637