
6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)chroman 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1002727-88-9
:
Formula:C₁₅H₂₁BO₃
MDL Number:MFCD12028565
:260.14 g/mol
Melting Pt:72…74.5 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together