
1-Tritylimidazole-2-carbaldehyde
Catalog # APOSOR1652-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:338.400 g/mol
Melting Pt:189…193 °C
:GHS07
Storage Temperature:Ambient
CAS Number:67478-50-6
Formula:C₂₃H₁₈N₂O
MDL Number:MFCD03453110