
1-[2-(Benzhydryloxy)ethyl]-1,4-diazepane
Catalog # APOSOR1581-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:310.44 g/mol
Boiling Pt:183 °C/0.5mmHg
:GHS07
Storage Temperature:Ambient
CAS Number:150557-09-8
Formula:C₂₀H₂₆N₂O
MDL Number:MFCD00729027