Zadání objednávky
Czech Republic
ContactUsLinkComponent
(S)-3-Hydroxypyrrolidine hydrochloride
Irritant
(S)-3-Hydroxypyrrolidine hydrochloride
Catalog # APOSOR15682-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
(S)-3-Hydroxypyrrolidine hydrochloride
Catalog # APOSOR15682-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Typ balení:
    Bottle
  • Bal.:
    5 g
  • Sekvence:
    3173

Specifications

About this item

 :
 :123.58 g/mol
Boiling Pt:108…110 °C
Melting Pt:106…107 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:122536-94-1
Formula:C₄H₁₀ClNO
MDL Number:MFCD00272298