1-(4-Bromophenyl)-2-(methylsulphonyl)-1-ethanone
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:500576-33-0
:
Formula:C₉H₉BrO₃S
MDL Number:MFCD09864834
:277.14 g/mol
Melting Pt:152…154 °C
Storage Temperature:Ambient
Frequently Bought Together