Benzyl-4-[5-chloro-1-(2-ethoxy-2-oxoethyl)-6-oxo-1,6-dihydro-4-pyridazinyl]tetrahydro-1(2H)-pyrazine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1000018-18-7
:
Formula:C₂₀H₂₃ClN₄O₅
MDL Number:MFCD09864828
:434.88 g/mol
Melting Pt:116…117 °C
Storage Temperature:Ambient
Frequently Bought Together