4-Hydroxy-6-(2-methoxyphenyl)-2-phenyl-5-pyrimidinecarbonitrile
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:477853-96-6
:
Formula:C₁₈H₁₃N₃O₂
MDL Number:MFCD00794104
:303.32 g/mol
Melting Pt:277…280 °C
Storage Temperature:Ambient
Frequently Bought Together